To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Immunoglobulin L
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:IgL
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Epitope:Light Chain
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-80/106
- Purification:Purified by Protein A
- Cat. no.:10393-908
- Supplier No.:BS-3801R-CY5
Spécifications
A propos de cet article
All five immunoglobulin classes share the same basic four polypeptide chain structure of two heavy-chains and two light chains. There are five heavy chain types, and two light-chain types (Kappa and Lambda) both having a molecular weight of 22.5kDa. Any heavy-chain type can associate with either light-chain type, but on any immunoglobulin molecule both light-chains are of the same type. Kappa and Lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Human