To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ribonucleic Acid Export 1
- Clonality:Polyclonal
- Gene ID:8480
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RAE1
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:mRNA-associated protein MRNP 41|MIG14|MGC126076|OTTHUMP00000166110|RP4-800J21.2|mRNA-binding protein|dJ481F12.3|mRNA export protein|migration-inducing gene 14|MGC126077|MRNP41|HGNC:9828|RAE1|Mnrp41|S.pombe) homolog|homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer)|MGC117333|FLJ30608|41-kD|dJ800J21.1|RAE1 RNA export 1 homolog (S. pombe) |RAE1 (RNA export 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10393-842
- Supplier No.:BS-3791R-A555
Spécifications
A propos de cet article
Mutations in Rae1 and Gle2(Saccharomyces cerevisae homolog) genes have been shown to result in accumulation of poly(A)-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Depletion of Rae1 from extracts or cells severely inhibits mitotic spindle assembly. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAE1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat