To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 4
- Clonality:Polyclonal
- Gene ID:3805
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:KIR2DL4
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:KIR103|killer cell immunoglobulin-like receptor 2DL4|natural killer cell inhibitory receptor|killer Ig receptor|killer cell inhibitory receptor 103AS|MHC class I NK cell receptor KIR103AS|CD158 antigen-like family member D|NK cell receptor|KIR103AS|CD158D|G9P|KIR-103AS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/377
- Purification:Purified by Protein A
- Cat. no.:10343-040
- Supplier No.:BS-2643R-CY7
Spécifications
A propos de cet article
Receptor on natural killer (NK) cells for HLA-C alleles. Inhibits the activity of NK cells thus preventing cell lysis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KIR2DL4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human