To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 15 Receptor Alpha
- Clonality:Polyclonal
- Gene ID:3601
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:IL15RA
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CD215|IL-15RA|IL-15 receptor subunit alpha|Interleukin-15 receptor subunit alpha
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/267
- Purification:Purified by Protein A
- Cat. no.:10342-594
- Supplier No.:BS-2605R-A488
Spécifications
A propos de cet article
IL15RA is a cytokine receptor that specifically binds IL15 with high affinity. It shares two subunits with the receptor of IL2, the IL2R beta and IL2R gamma chains. This forms the basis of many overlapping biological activities of IL15 and IL2. The IL2 receptor requires an additional IL2-specific alpha subunit for high affinity IL2 binding. This protein is structurally related to IL2R alpha, but is capable of binding IL15 with high affinity independent of other subunits, which suggests the distinct roles between IL15 and IL2. This receptor is reported to enhance cell proliferation and expression of apoptosis inhibitor BCL2L1/BCL2-XL and BCL2. There are nine different isoforms.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL15RA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat