To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Apelin
- Clonality:Polyclonal
- Gene ID:8862
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:APLN
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:XNPEP2|APEL
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10336-082
- Supplier No.:BS-2425R-A555
Spécifications
A propos de cet article
Apelin is a neuropeptide expressed in the supraoptic and paraventricular nuclei and is an endogenous ligand for APJ, a G protein-coupled orphan receptor which is an alternative coreceptor with CD4 for HIV-1. Apelin and APJ are ubiquitously expressed in peripheral tissues, with highest levels reported for heart and lungs, as well several regions within the central nervous system. The actions of apelin remain largely unknown: Apelin inhibits HIV-1 entry in cells coexpressing CD4 and APJ; the oral intake of Apelin in colostrum and breast milk could have a role in the modulation of the immune responses in neonates; more recent studies have also indicated a role for Apelin in the central control of body fluid homeostasis, by influencing AVP release and drinking behavior. In the cardiovascular system several actions of Apelin have been described, including endothelium-dependent vasodilatation, vasoconstriction through direct action on smooth muscle and positive inotropism.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APLN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat