To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 1 Receptor-like 1
- Clonality:Polyclonal
- Gene ID:9173
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:IL1RL1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Lymphocyte antigen 84|T1/ST2|Fit-1|ST2|Ly84|DER4|Interleukin-1 receptor-like 1|St2-rs1|Ste2|ST2L|T1|St2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:19-36/556
- Purification:Purified by Protein A
- Cat. no.:10334-066
- Supplier No.:BS-2382R-PE
Spécifications
A propos de cet article
Receptor for interleukin-33 (IL-33), its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. Possibly involved in helper T-cell function.
Recommended Dilutions: Flow Cytometry: 1:20-100
Type: Primary
Antigen: IL1RL1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat