To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:NAD(P)H Dehydrogenase, Quinone 1
- Clonality:Polyclonal
- Gene ID:1728
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NQO1
- Conjugation:HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:NMORI|NMOR1|dioxin-ind|diaphorase (NADH/NADPH)|quinone 1|NQO1|diaphorase (NADH/NADPH) (cytochrome b-5 reductase)|NAD(P)H:quinone oxireductase|DIA4|azoreductase|DTD|NAD(P)H:menadione oxidoreductase 1|dioxin-inducible 1|NAD(P)H menadione oxidoreductase 1|DHQU|NAD(P)H dehydrogenase|dioxin-inducible|QR1|NAD(P)H:Quinone acceptor oxidoreductase type 1|DT-diaphorase|diaphorase-4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:240-274/274
- Purification:Purified by Protein A
- Cat. no.:10325-670
- Supplier No.:BS-2184R-HRP
Spécifications
A propos de cet article
The enzyme apparently serves as a quinone reductase in connection with conjugation reactions of hydroquinons involved in detoxification pathways as well as in biosynthetic processes such as the vitamin K-dependent gamma-carboxylation of glutamate residues in prothrombin synthesis.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat