To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Growth Associated Protein 43
- Clonality:Polyclonal
- Gene ID:14432
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GAP43
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:axonal membrane protein GAP-43|growth-associated protein 43|neuron growth-associated protein 43 protein F1|neuromodulin|nerve growth-related peptide GAP43|calmodulin-binding protein P-57|neural phosphoprotein B-50|PP46|B-50
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1-50/227
- Purification:Purified by Protein A
- Cat. no.:10232-224
- Supplier No.:BS-0154R-A555
Spécifications
A propos de cet article
This protein is associated with nerve growth. It is a major component of the motile "growth cones" that form the tips of elongating axons. Plays a role in axonal and dendritic filopodia induction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat