To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Actin Beta
- Clonality:Polyclonal
- Gene ID:60
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Antigen symbol:ACTB
- Conjugation:Cy5.5®
- ImmunoFluorescence:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1-50/375
- Purification:Purified by Protein A
- Cat. no.:10230-332
- Supplier No.:BS-0061R-CY5.5
Spécifications
A propos de cet article
Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells.
Recommended Dilutions: IF(ICC)(1:50-200
Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Pig