To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Caspase 4 And Caspase 13
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:CASP4 + CASP 13
- Conjugation:Cy5.5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Caspase 13 subunit 2|CASP-13|apoptosis related cysteine|Caspase-13|ERICE.|CASPD_BOVIN|Caspase 4|Caspase 13 subunit 1|CASP4|Caspase 4 subunit 1|Evolutionary related interleukin-1-beta-converting enzyme
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10230-132
- Supplier No.:BS-0053R-CY5.5
Spécifications
A propos de cet article
Caspase-13 (also designated Evolutionarily related interleukin-1beta converting enzyme ; ERICE) involved in the activation cascade of caspases responsible for apoptosis execution. Might function by either activating some proteins required for cell death or inactivating proteins necessary for cell survival. [subunit] Heterodimer of a small and a large subunit. The two subunits are derived from the precursor sequence by an autocatalytic mechanism or by cleavage by Caspase-8. Mainly expressed in peripheral blood lymphocytes, spleen and placenta. Belongs to peptidase family C14.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CASP4 + CASP 13
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Bovine