To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Melatonin Receptor 1A
- Clonality:Polyclonal
- Gene ID:4543
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Antigen symbol:MTNR1A
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MEL-1A-R|MEL1A|melatonin receptor type 1A|melatonin 1|melatonin 1A|MT1|mel1a receptor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:209-242/350
- Purification:Purified by Protein A
- Cat. no.:10229-576
- Supplier No.:BS-0027R-CY3
Spécifications
A propos de cet article
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: MTNR1A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine