To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Presenilin 1
- Clonality:Polyclonal
- Gene ID:5663
- Host:Mouse
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat
- Antigen symbol:PSEN1
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:FAD|Alzheimer Disease 3|Homo Sapiens Clone CC44 Senilin 1|Senilin 1|PS1|PSNL1|AD3|S182 Protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10229-530
- Supplier No.:BS-0025M-CY5
Spécifications
A propos de cet article
Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1;PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor, such that they either directly regulate gamma-secretase activity or themselves are protease enzymes. Three alternative splice variants of PSEN1 have been identified.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PSEN1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat, Rabbit