Graphic Display Sound Meter, Sper Scientific
Fournisseur: Sper Scientific
Certificats
A propos de cet article
Meters feature a large bar graph display, A and C decibel frequency weighting scales, min./max., fast or slow response, AC or DC output, auto-power-off, and low battery indicator
Meets ANSI S1.4 Type 2 and IEC 61672-1 Class 2. N.I.S.T. traceable certificate of calibration available, or calibrate in compliance with OSHA anytime using optional Acoustical Calibrator 850016. Covers 30 ~ 130 dB in the A and C scales with a resolution of 0.1 dB, with an accuracy of ±1.4 dB (ref 94 dB@1KHz).
Ordering information: Supplied ready to use with a windscreen, 9V battery, instructions, and a foam lined hard shell carrying case. The Certified meter includes a N.I.S.T. traceable certificate of compliance. All units include a manufacturer's five-year warranty.
Spécifications
- Catalog No:
- 10025-388
- 10025-390
- Dimensions:
- 258 × 55 × 25 mm (101/4 × 21/4 × 1")
- 258 × 55 × 25 mm (101/4 × 21/4 × 1")
- Weight:
- 185 g (6.5 oz.)
- 185 g (6.5 oz.)