ContactUsLinkComponent
Indoor Air Quality Wall Monitor, Sper Scientific
Indoor Air Quality Wall Monitor, Sper Scientific
# de catalogue 10025-402
Fournisseur:  Sper Scientific
Indoor Air Quality Wall Monitor
Indoor Air Quality Wall Monitor, Sper Scientific
# de catalogue 10025-402
Fournisseur:  Sper Scientific
Spécifications
  • Description:
    IAQ Wall Monitor 
  • Cat. no.:
    10025-402
  • Dimensions:
    304×216×51 mm (12 × 81/2 × 2")
  • Weight:
    1315 g (2.9 lbs.)
  • Supplier No.:
    800051
Spécifications
A propos de cet article
Visible across a large room
This instrument is ideal for monitoring indoor air quality in crowded public spaces with potentially high levels of CO2 (carbon dioxide), such as offices, factories, classrooms, hospitals, and hotels. Displays readings on a bright green LED display with 2½" high CO2 digits. Also indicates general CO2 levels with green (good), yellow (fair), and red (poor) LED lights. User may set a maximum CO2 audible alarm and adjust temperature and relative humidity readings to match any standard. Automatic Baseline Calibration (ABC) and maintenance free NDIR (non-dispersive infrared) CO2 sensor ensure long-term accuracy, stability, and reliability.
Continually monitors the primary IAQ (Indoor Air Quality) parameters, including Air Temperature, Relative Humidity, and CO2.
Ordering information: Supplied with an AC adaptor and manufacturer's five-year warranty. Unit will stand on a desk-top or can be wall mounted.
Customers Who Bought This Also Bought