Indoor Air Quality Monitor, Sper Scientific
# de catalogue 10025-396
Fournisseur: Sper Scientific
Spécifications
- Description:IAQ Monitor
- Cat. no.:10025-396
- Dimensions:108×115×25 mm (4½ × 4¼ × 1")
- Weight:170 g (6 oz.)
- Supplier No.:800048
Spécifications
A propos de cet article
Large display continually monitors and records the primary IAQ (Indoor Air Quality) parameters, including Air Temperature, Relative Humidity, and CO2.
The Indoor Air Quality (IAQ) monitor has a large, easy-to-read display that continually presents primary IAQ parameters (Air Temperature, Relative Humidity, CO2). The unit recalls min./max. CO2 levels and provides visual alarms for maximum CO2 threshold. Automatic Baseline Calibration (ABC) and maintenance-free NDIR (non-dispersive infrared) CO2 sensor ensure long-term accuracy, stability, and reliability.
Ordering information: Supplied with an AC adaptor and manufacturer's five-year warranty. Unit will stand on a desk-top or can be wall mounted.
Customers Who Bought This Also Bought