Heat Stress Monitor, Sper Scientific
# de catalogue 10025-392
Fournisseur: Sper Scientific
Spécifications
- Description:Heat Stress Monitor
- Cat. no.:10025-392
- Dimensions:108×115×25 mm (41/2 × 41/4 × 1")
- Weight:141 g (5 oz.)
- Supplier No.:800034
Spécifications
A propos de cet article
The unit features a large display that continually monitors all heat stress indicators.
This heat stress monitor has a large display that continually monitors temperature, relative humidity, and all heat stress indicators. These include Air Temperature, Relative Humidity, Heat Index, Dew Point, and Wet Bulb. The unit recalls min./max. temperature and provides visual alarms for user-selectable hi/lo temperature and relative humidity thresholds.
Ordering information: Includes four AAA batteries and stand for a desk-top or wall mount. Unit also includes a five-year manufacturer's warranty.
Customers Who Bought This Also Bought