MicroOne Bacteria Removing Wipes, Vileda Professional-FHP
Fournisseur: Vileda Professional - FHP
Certificats
A propos de cet article
MicroOne has proven performance of 99.9% bacterial removal.
MicroOne comes packed in re-sealable plastic bags that fit perfectly in our 6 L bucket for maximum hygiene and handling. The bag is excellent for use in pre-prepared cleaning. Use 500 ml per 50/cloths for general cleaning, and 600 ml per 50/cloths for disinfecting. Use once and throw away. Significantly reduces the risk of cross-contamination.
Customers Who Bought This Also Bought