About this item
Merit™ operating scissors, also known as surgical scissors are a versatile surgical instrument
Merit™ instruments are office-grade line of products used for routine and in-office minor procedures.
These high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
They are used for general, multipurpose cutting and dissecting. Sharp operating scissors cut tougher tissue and are frequently used for dissecting, whereas blunt operating scissors are more often used for cutting materials such as sutures, tubing, or gauze.