Baseline® Clinical Tuning Forks
For testing hearing and sensation.
- Available Individually or in sets
- Variable Cycle option
- Lightweight alluminum construction
This standard tool is designed to evaluate hearing and vibratory sensation in the industry. The clinical grade set includes six forks: 128, 256, 512, 1024, 2048, and 4096 cps.
Customers Who Bought This Also Bought