Sims Scissors, OR Grade, Sklar
Fournisseur: Sklar
Certificats
A propos de cet article
Sims scissors are finger ring instruments, that feature long blades for deep tissue dissection.
Ordering information: Available in curved tip style.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.