3B Scientific® Deluxe Head
# de catalogue 470119-052
Fournisseur: American 3B Scientific
Spécifications
- Description:Deluxe Head
- Height:23 cm
- Cat. no.:470119-052
- Length:20 cm
- Width:24 cm
- Supplier No.:C09/1
- No. of Parts:6
Spécifications
A propos de cet article
This six-part head is mounted on a base and features a removable four-part brain half with arteries.
- 3 Year Manufacturers Warranty
- Includes 3B Smart Anatomy
The eyeball with optic nerve is also removable and one side of the high quality model exposes the nose, mouth cavity, pharynx, occiput and skull base. On a removable base for easy display in a classroom or doctor's office.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought