Permafrost Adhesion Economy Frosted Microscope Slides, Thermo Scientific
Fournisseur: Erie Scientific
Certificats
A propos de cet article
Permafrost Adhesion Slides are designed for tissue adhesion in routine clinical applications
All Permafrost Adhesion slides are pre-cleaned, measure 25x75x1mm, feature smoothly ground edges, and are manufactured from white Swiss glass. Slides are packed approximately 72 per pack, 20 packs per case.
Permafrost Adhesion slides feature a ¾” frosted end with two "a" markings at the bottom of each slide. They are available in nine different colors.
Customers Who Bought This Also Bought