Bouin's Solution, Azer Scientific
Fournisseur: VWR International
Certificats
A propos de cet article
Bouin's solution is a saturated picric acid-formalin fixative that is useful for preserving delicate tissues and gastrointestinal tract specimens.
Packaging: Dispending spigots are included with cube containers.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.