OptiTherm® Reaction Blocks, Round Bottom, Chemglass
Supplier: Chemglass
Certificates
About this item
OptiTherm® blocks provides a safe and cost effective solution for replacing your existing oil baths and heating mantles.
The innovative PTFE shield on the unit keeps the exterior surface area at a temperature that is safe to touch.
Customers Who Bought This Also Bought