Bottle Cap Assembly, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Bottle cap for GL-45 and GPI-38 threads that permits standard solvent or media bottles to be used as an HPLC reservoir.
- Caps are made from polypropylene
- Inserts are made from tefzel
Complete cap assemblies consists of the cap, tefzel insert, two plugs for the 1/8" tubing openings and a plug for the opening. Caps have two holes to accomodate 1/8" tubing and one hole that accepts a standard luer inner joint.
Customers Who Bought This Also Bought