SMZ-171 Stereo Microscope, Motic
Fournisseur: Motic
Certificats
A propos de cet article
Flexible stereo microscopes for advanced research applications in biomedicine and material science. The 6.7:1 Greenough zoom delivers a standard magnification of 7.5x - 50x with 110 mm working distance in a FOV23mm.
- Eyepieces feature Widefield N-WF10X/23 mm with diopter adjustment
Delivery: Supplied with the following: Stands, Stage Accessories, Micrometer Reticles, Additional Illumination, Polarizing Equipment, Darkfield Equipment, Barlow Lenses. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative.
Spécifications
- Catalog No:
- CA10769-598
- CA10769-604
- CA10769-600
- CA10769-602
- Magnification:
- 7.5x to 50x
- 7.5x to 50x
- 7.5x to 50x
- 7.5x to 50x
- Illumination:
- None or LED (optional)
- None or LED (optional)
- None or LED (optional)
- None or LED (optional)
- Electrical:
- 100–240V
- 100–240V
- 100–240V
- 100–240V