ContactUsLinkComponent
Foam Biopsy Pads, Globe Scientific
Foam Biopsy Pads, Globe Scientific
# de catalogue 10022-166
Fournisseur:  Globe Scientific
Biopsy Pad for Cassettes, Foam, Blue, 30.2mm x 25.4mm x 2mm
Foam Biopsy Pads, Globe Scientific
# de catalogue 10022-166
Fournisseur:  Globe Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Blue
  • Description:
    Foam Biopsy Pads
  • Cat. no.:
    10022-166
  • Supplier No.:
    190195
Spécifications
A propos de cet article
The Globe Scientific foam biopsy pads are used to hold biopsies in place and prevent their loss during processing
They are produced from a special polyester urethane foam, which is verified for consistency throughout in order to ensure optimum solvent flow. Simply sandwich the biopsy sample between two pads and insert into a cassette for processing.
Customers Who Bought This Also Bought