Refrigerated and Heated Bath/Circulators, Caron Products
Fournisseur: Caron Products
Certificats
A propos de cet article
This refrigerated bath-circulator is designed to strike a special balance between strong cooling/heating/circulation capacity and compact cube dimensions, making it ideal for use with electron microscopes, rotary evaporators, lasers and spectrophotometers.
- Low unit height fits under counters or in small cabinets
- Integral control head design saves space
- Entire unit takes up less than 1.5 cu. ft. of space
- All wetted surfaces are chemical resistant stainless steel
- Sealed control panel
- Future-proof natural refrigerant for easy long-term servicing
Features quick-connect inlet and outlet fittings with built-in over and under temperature tracking alarms and low liquid level shutoff. It has a big bright LED display, with simple push button menu-driven programming.
Spécifications
- Catalog No:
- 76355-514
- 76355-516
- Heating capacity:
- 0.75 kW
- 0.75 kW
- Cooling capacity:
- 300W (1025 BTU/Hr.) @ 20 °C
- 300W (1025 BTU/Hr.) @ 20 °C
- Pump flow rate:
- 6.5 LPM at 0' head (External Pumping)
- 6.5 LPM at 0' head (External Pumping)
- Temperature control:
- ±0.1 °C
- ±0.1 °C
- Refrigerant:
- R290
- R290
- Alarm:
- Over–temperature and low liquid level (Safety Alarms)
- Over–temperature and low liquid level (Safety Alarms)
- W×D×H:
- 35.6×34.3×35.6 cm (14×13.5×14")
- 35.6×34.3×35.6 cm (14×13.5×14")
Spécifications
Informations connexes
Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product.
PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.
PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.