Accessories for KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Test fabric is useful for testing dyes on a variety of fabrics
Three-foot length of fabric four inches wide features 8 strips of different fibers woven side by side. Filament acetate, bleached cotton, spun polyamide type 66, polyester disperse dyeable, spun polyacrylic, spun silk, spun viscose, and worsted wool.
Accessory for Microscale Williamson kit.
Customers Who Bought This Also Bought