Spécifications
- Dimensions:6×9¹/₄"
- Description:4-wall sample bags with document pouch for chain of custody transfers
- Cat. no.:89400-296
- Supplier No.:SPECI-CC
Spécifications
A propos de cet article
Flap, double pocket bag with two separately sealed inside pockets.
- Includes adhesive closure
- The flap, when folded over, seals in both pockets
- It keeps the requisition and specimen sealed for forensic testing
The bottom line of adhesive seals the inside pocket liquid tight. Each bag includes a pre-inserted 3×3" absorbent sheet which can absorb up to 60 ml of liquid.