3B Scientific® Dual Sex Torso With Muscular Arm
# de catalogue 470119-030
Fournisseur: American 3B Scientific
Spécifications
- Description:Dual Sex Torso With Muscular Arm
- No. of Keyed Items:786
- Cat. no.:470119-030
- Supplier No.:B42
- No. of Parts:33
Spécifications
A propos de cet article
The high-quality human torso represents the complete anatomy of the human upper body, especially suitable for the high standards of medical school.
- Mounted On Stand For Transportability
- Includes Male And Female Inserts
- 3 Year Manufacturers Warranty
- Includes 3B Smart Anatomy
The versatility of this aesthetically designed human torso model is rounded off by the exact representation of the internal organs. The right half of the torso shows the skin. The left half the superficial and deeper muscles with nerves, vessels and bony structures thanks to a removable six-part muscle arm.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought