MINUM-WARE®, Neoprene Filter Adapters, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Microscale neoprene filter adapters for use with MW-122 filtering flasks.
Adapters may be nested to permit use of funnels with a stem diameter as small as 6 mm O.Ø.
Customers Who Bought This Also Bought