Cover Slips, Quartz, Chemglass
# de catalogue 80063-860
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Quartz cover slips, square
- L×W:19×19 mm
- Thickness:0.25 mm
- Cat. no.:80063-860
- Supplier No.:CGQ-0660-01
Spécifications
A propos de cet article
These cover slips made of clear quartz are ultra-thin, ground and polished.
Customers Who Bought This Also Bought