ECO Funnels with HPLC Waste Hook Adapter, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
ECO funnels are supplied with an HPLC waste hook adapter that connects to the side of the funnel, eliminating the danger of dangling waste lines into bottles and containers.
Customers Who Bought This Also Bought