96-Well Preparation Rack with Cover, Bio Plas
Fournisseur: Bio Plas
Certificats
A propos de cet article
High quality
- Designed for preparation and storage
- Racks for use with a variety of microcentrifuge tubes
Manufactured of polypropylene, these 96-well (8x12 format) racks are suitable for autoclaving. These one sided racks are suitable for 0.2 ml PCR tubes with strip caps. The snap-on cover stays in place during transportation and storage. Racks are able to be securely stacked.
Made in the USA.
Customers Who Bought This Also Bought