Refractive Index Liquids, Series AAA
# de catalogue 47735-584
Fournisseur: Cargille Laboratories
Numéro CAS:
Spécifications
- Description:Series AAA-x, standard nD 1.300 - 1.395
- Size:7 ml (0.25 oz.)
- Supplier No.:1803X
Spécifications
A propos de cet article
Refractive index liquids have a broad range of applications in diversified fields.
- Slightly volatile
- Colorless flurocarbon formulation
Adjustment: ±0.0002
Choose from Range nD 1.300-1.395
Ordering information: Required refractive index has to be specified with order.
Customers Who Bought This Also Bought