Spécifications
- Description:Enzyme Catalyzed Reactions Kit
- No. of Lab Groups:10
- Cat. no.:470218-354
- Supplier No.:IS3502
Spécifications
A propos de cet article
Living organisms require enzymes to regulate the chemical reactions necessary to maintain life.
- All Necessary Chemicals Provided
- Three Enzymes
- Teacher's Guide and Student Material Provided
The activities in this kit introduce students to the concept of enzymes. Students perform a series of four experiments involving invertase, amylase, and papain to demonstrate different enzyme functions, activities, and specificities.
This kit contains enough material for ten groups.