VWR® Patient ID Labels
Catalog # 89097-834
Supplier: VWR International
Specifications
- Color:Blank White
- Description:VWR® patient ID labels
- Dimensions:25.4×76.2 mm (1×3")
- Cat. no.:89097-834
- Packaging:1000 labels/roll
Specifications
About this item
Smudge-proof flexo labels feature a removeable adhesive for patient ID labeling.
- Multiple lines for identification
- Smudge-proof
- Carrier width is 82.5 mm
All labels allow the user to dial in two lines of information with up to eight characters per line to be printed. Available characters include two Gregorian dates for printing expiration and autoclave dating with an additional band for load and sterilizer numbers.
Customers Who Bought This Also Bought