3B Scientific® Vegetative Nervous Chart
# de catalogue 470120-182
Fournisseur: American 3B Scientific
Spécifications
- Length:50 cm
- Width:67 cm
- Description:Vegetative Nervous Chart
- Cat. no.:470120-182
- Finish:Laminated
- Supplier No.:VR1610L
Spécifications
A propos de cet article
This vegetative nervous system chart is a must for any classroom or doctor's office.
This vegetative nervous system chart is a must for any classroom or doctor's office.
The function of this part of the nervous system is explained on this poster, but the role the vegetative nervous system plays in the function of each organ is detailed.
Customers Who Bought This Also Bought