Baseline® Three Point Descriminator
# de catalogue 470119-640
Fournisseur: American 3B Scientific
Spécifications
- Description:Three Point Descriminator
- Cat. no.:470119-640
- Supplier No.:W54068
Spécifications
A propos de cet article
Accurately test two point sensitivity.
- Sliding point measured in mm
- Switch between one and two point touch easily
Evaluate cutaneous sensitivity and touch threshold. Plastic points minimize influence of temperature. Two point discriminator has third point that permits alternating between single and double point stimulation without changing setting, measures up to 14 cm discrimination.
Customers Who Bought This Also Bought