Tactile Monofilament Evaluator, Foot Set
# de catalogue 470119-636
Fournisseur: American 3B Scientific
Spécifications
- Description:Tactile Monofilament Evaluator, Foot Set, 6 Pieces
- Cat. no.:470119-636
- Supplier No.:W54066
Spécifications
A propos de cet article
The baseline tactile monofilament evaluator, foot set, measures cutaneous sensory perception threshold of the feet. Used to test sensation levels, and obtain objective data for accurate reporting on the status of diminishing or retuning sensibility.
Features 2.83, 3.61, 4.31, 4.56, 5.07 and 6.65 sized monofilaments.
Customers Who Bought This Also Bought