VWR® Preprinted Flammable Warning Tape
# de catalogue 10159-644
Fournisseur: VWR International
Spécifications
- Dimensions:12.7 mm x 20.3 m
- Color:White with Red
- Cat. no.:10159-644
Spécifications
A propos de cet article
VWR® Preprinted Flammable Warning Tape is imprinted with red ink on white tape and can be used on virtually any surface where visual warning is required.
- Durable
- Self-adhesive design
- No residue
This durable, self-adhesive tape can withstand temperatures from -23 to 121°C (-10 to 250°F). The adhesive backing will peel off easily and leaves no residue.
Customers Who Bought This Also Bought