Spécifications
- Color:Black
- Description:VWR® Black Two-Line Labeling Gun
- Cat. no.:10123-976
Spécifications
A propos de cet article
This two-line labeling gun features an ergonomic handle, reinforced ABS body with steel internal components, easy drop-in label loading, a simple snap-in/pop-out ink roller, and a self-align feed mechanism that provides jam-resistant operation.
- Autoclavable adhesive withstands extremely high temperatures for up to 20 minutes
- Easy to replace ink roller
All labels allow the user to dial in two lines of information, with up to eight characters per line to be printed
The accessory easy pop-in/out ink roller is a replacement roller for the VWR® Black Labeling Gun. It is recommended to change the ink roller after using two sleeves of labels.
Available characters include two Gregorian dates for printing expiration and autoclave dating with an additional band for load and sterilizer numbers. All gun labels are printed on an autoclavable adhesive that withstands temperatures up to 132°C (270°F) for 20 minutes.