Baseline® Tactile Monofilaments Sensory Evaluators
# de catalogue 470119-634
Fournisseur: American 3B Scientific
Spécifications
- Description:Tactile Monofilaments Sensory Evaluators Set, 20 Pieces
- Cat. no.:470119-634
- Supplier No.:W54065
Spécifications
A propos de cet article
Quantify Sensation Levels.
- Units measure cutaneous sensory perception thresholds
- Each monofilament represents a unique amount of force
- Twenty monofilaments included
- Protective case included
These evaluators help test sensation levels and obtain objective data for accurate reporting on the status of diminishing or retuning sensibility. They measure cutaneous sensory perception thresholds. Each monofilament represents a unique amount of force ranging from 1.65 to 6.65 in size and .008 to 300 grams of force. The full set stores in a protective case.
Customers Who Bought This Also Bought