Baseline® Wartenburg Pinwheel
# de catalogue 470119-632
Fournisseur: American 3B Scientific
Spécifications
- Description:Wartenburg Pinwheel
- Cat. no.:470119-632
- Supplier No.:W54063
Spécifications
A propos de cet article
Examine Cutaneous Perception.
- Stainless Steel Handle
- Textured Grip
- Rotating Spur
It's designed for the evaluation of gross cutaneous sensory perception and pain perception responses.
The pinwheel has an overall length of 19 cm (71/2").
Customers Who Bought This Also Bought