ContactUsLinkComponent
Ward's® Chemistry Safer Sodium Demonstration
Catalogue # 470118-182
Fournisseur:  Ward's Science
undefined
Ward's® Chemistry Safer Sodium Demonstration
Catalogue # 470118-182
Fournisseur:  Ward's Science
Supplier Number:  AD-CC0266-8KIT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Ward's® Chemistry Safer Sodium Demonstration
  • No. of Lab Groups:
    5
  • Cat. no.:
    470118-182
  • Supplier No.:
    AD-CC0266-8KIT
Spécifications
À propos de cet article
This demonstration is a safer way to show the reaction between water and alkali metals.
  • A Safer Way to Show the Reaction Between Water and Alkali Metals
  • Demonstration Displays Density, Chemical Reactions, and pH Indicators
  • Cost and Time Efficient
  • For Grades 5-12
  • Time Requirement: 10 Minutes
In this demonstration, students mix two immiscible liquids, mineral oil and water, in a graduated cylinder. A small amount of sodium in the mixture permeates through the mineral oil and react with the two liquids on the bottom.
The reaction, as gas bubbles, form and rise to the mineral oil layer and then drop back down to the water and react yet again. A pH indicator to the water layer displays the production of a base when sodium and water react.
Certifications: Meets NSES for grades five through 12.
Ordering information: This kit includes sodium, phenolphthalein, and mineral oil. Water, graduated cylinders, and forceps are required but not provided.