Isometric Goniometers
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
These clear plastic goniometers permit the observation of joint's axis of motion and range of motion.
- Calibrated in 3 scales
- 360 degree scale
The 360º head has three scales calibrated for use with the ISOM (International Standards of Measurement) system.
Linear scale in inches and centimeters.
Customers Who Bought This Also Bought