Baseline Pinch Gauge, AMERICAN 3B SCIENTIFIC SE
# de catalogue 470119-608
Fournisseur: American 3B Scientific
Spécifications
- Description:Baseline Pinch Gauge, Red
- Cat. no.:470119-608
- Supplier No.:W50181
Spécifications
A propos de cet article
The baseline gauge measures pinch strength, with reliable and repeatable readings.
Measures tip, key and palmar pinch strength of healthy hands,
Consistent results with the published Mathiowetz studies,
Indicator remains at the subject's maximum reading until reset,
Strength readings in pounds (60 lbs.) and kilograms (27 kg),
Lightweight anodized aluminum with wrist strap and carrying case.
Customers Who Bought This Also Bought