Baseline Goniometer
# de catalogue 470119-604
Fournisseur: American 3B Scientific
Spécifications
- Description:Baseline Goniometer
- Cat. no.:470119-604
- Supplier No.:W50179
Spécifications
A propos de cet article
This stainless steel goniometer measures the range of motion of finger joints (metacarpophalangeal and interphalangeal) and other small joints including toes.
The goniometer's head has two opposing 180º scales marked in 5º increments. A linear scale in inches and centimeters is on arm of goniometer.
Customers Who Bought This Also Bought