Pinch Gauge, AMERICAN 3B SCIENTIFIC SE
# de catalogue 470119-598
Fournisseur: American 3B Scientific
Spécifications
- Description:Pinch Gauge
- Cat. no.:470119-598
- Supplier No.:W50176
Spécifications
A propos de cet article
This 6.35 cm (2,5") diameter head hydraulic pinch gauge offers accurate and repeatable pinch strength measurements. The hydraulic system assures convenience, product reliability, measurement accuracy and repeatability. For all pinch test: Tip, key and palmar.
Registers up to 50 lbs. (22.7 kg)
Customers Who Bought This Also Bought